Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002511.3 | NP_002502.2 | 1, 2 | 567 | ||
GenBank mRNA | BC095542.1 | - | 1, 2 | 427 | 7001999 | |
GU727635.1 | - | 1, 2 | 426 | |||
M73482.1 | - | 1, 2 | 565 |
Set Membership: |
![]() |