Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324307.1 | NP_001311236.1 | 3 | 278 | ||
NM_001324308.1 | NP_001311237.1 | 2 | 203 | |||
NM_002511.3 | NP_002502.2 | 2 | 704 | |||
XM_011535846.1 | XP_011534148.1 | 4 | 334 | |||
XM_017010901.1 | XP_016866390.1 | 2 | 303 | |||
XM_017010902.1 | XP_016866391.1 | 2 | 240 | |||
GenBank mRNA | BC095542.1 | - | 2 | 564 | 7001999 | |
DB462076.1 | - | 4 | 368 | |||
GU727635.1 | - | 2 | 563 | |||
M73482.1 | - | 2 | 702 |
Set Membership: |
![]() |