Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002522.3 | NP_002513.2 | 5 | 1394 | ||
GenBank mRNA | AK124701.1 | - | 3 | 1837 | ||
AK127335.1 | - | 5 | 1394 | |||
AL119012.1 | - | 1 | 96 | |||
BC089441.1 | - | 5 | 1378 | 30555728 | ||
U61849.1 | - | 5 | 1368 |