Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC11A2 SIRNA Assays ›
Gene Symbol:
SLC11A2
Gene Name:
solute carrier family 11 member 2
Gene Aliases:
AHMIO1, DCT1, DMT1, NRAMP2
Chromosome Location:
Chr.12: 50952263 - 51028335 on Build GRCh38
UniGene ID:
Hs.505545
Species:
Human
siRNA ID
HSS107307
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC11A2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000617.2
NP_000608.1
14
1512
NM_001174125.1
NP_001167596.1
14
1504
NM_001174126.1
NP_001167597.1
14
1512
NM_001174127.1
NP_001167598.1
14
1507
NM_001174128.1
NP_001167599.1
14
1507
NM_001174129.1
NP_001167600.1
14
1772
NM_001174130.1
NP_001167601.1
13
1487
NR_033421.1
14
1432
NR_033422.1
14
1507
XM_005268911.3
XP_005268968.1
14
1564
XM_005268912.4
XP_005268969.1
13
1689
XM_011538404.2
XP_011536706.1
14
1888
XM_011538405.2
XP_011536707.1
14
1496
XM_017019355.1
XP_016874844.1
14
1772
XM_017019356.1
XP_016874845.1
14
1702
GenBank mRNA
AB004857.1
-
14
1455
AB062284.1
-
14
1429
AF046997.1
-
14
1465
AF064484.1
-
14
1455
AJ493662.1
-
14
1504
AK094735.1
-
14
1772
AK128640.1
-
14
1404
AK296445.1
-
13
1466
AK315987.1
-
14
1418
AK316507.1
-
13
1443
AL832886.1
-
4
838
BC002592.2
-
14
1459
3160522
BC100014.1
-
14
1480
5300266
CD365137.1
-
5
716
KT455015.1
-
14
1468
L37347.1
-
11
1205
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
heme biosynthetic process
cobalt ion transport
copper ion transport
manganese ion transport
cellular iron ion homeostasis
learning or memory
response to iron ion
ferrous iron transport
lead ion transport
transferrin transport
copper ion transmembrane transport
dendrite morphogenesis
erythrocyte development
multicellular organismal iron ion homeostasis
cadmium ion transmembrane transport
ferrous iron import
manganese ion transmembrane transport
hydrogen ion transmembrane transport
ferrous iron transmembrane transport
copper ion transmembrane transporter activity
iron ion transmembrane transporter activity
manganese ion transmembrane transporter activity
protein binding
hydrogen ion transmembrane transporter activity
cadmium ion transmembrane transporter activity
cobalt ion transmembrane transporter activity
ferrous iron transmembrane transporter activity
lead ion transmembrane transporter activity
solute:proton symporter activity
ferrous iron uptake transmembrane transporter activity
transferrin transmembrane transporter activity
cadmium ion binding
transition metal ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0