Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101662.1 | NP_001095132.1 | 4 | 1112 | ||
NM_001242361.1 | NP_001229290.1 | 6 | 711 | |||
NM_002525.2 | NP_002516.2 | 6 | 1316 | |||
XM_005270903.1 | XP_005270960.1 | 4 | 459 | |||
XM_011541522.1 | XP_011539824.1 | 7 | 1305 | |||
XM_011541523.1 | XP_011539825.1 | 4 | 676 | |||
XM_011541525.2 | XP_011539827.1 | 6 | 774 | |||
XM_017001375.1 | XP_016856864.1 | 4 | 536 | |||
GenBank mRNA | AK055516.1 | - | 4 | 456 | ||
AK299271.1 | - | 4 | 559 | |||
AK302616.1 | - | 4 | 536 | |||
AK311631.1 | - | 4 | 506 | |||
AK312318.1 | - | 4 | 924 | |||
AY049784.1 | - | 4 | 827 | |||
AY360265.1 | - | 6 | 687 | |||
BC008775.2 | - | 4 | 886 | 3138712 | ||
U64898.1 | - | 4 | 884 | |||
X93207.1 | - | 6 | 1129 | |||
X93209.1 | - | 4 | 925 |