Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101662.1 | NP_001095132.1 | 1 | 341 | ||
NM_002525.2 | NP_002516.2 | 1 | 341 | |||
XM_011541522.1 | XP_011539824.1 | 1 | 167 | |||
GenBank mRNA | AK312318.1 | - | 1 | 153 | ||
AY049784.1 | - | 1 | 59 | |||
BC008775.2 | - | 1 | 118 | 3138712 | ||
DA245717.1 | - | 1 | 215 | |||
DA756064.1 | - | 1 | 341 | |||
U64898.1 | - | 1 | 116 | |||
X93207.1 | - | 1 | 154 | |||
X93209.1 | - | 1 | 154 |