Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTRK2 SIRNA Assays ›
Gene Symbol:
NTRK2
Gene Name:
neurotrophic receptor tyrosine kinase 2
Gene Aliases:
GP145-TrkB, TRKB, trk-B
Chromosome Location:
Chr.9: 84668368 - 85027070 on Build GRCh38
UniGene ID:
Hs.494312
Species:
Human
siRNA ID
HSS107338
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NTRK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007097.2
NP_001007098.1
9
1553
NM_001018064.2
NP_001018074.1
8
1344
NM_001018065.2
NP_001018075.1
9
1553
NM_001018066.2
NP_001018076.1
9
1553
NM_001291937.1
NP_001278866.1
11
1964
NM_006180.4
NP_006171.2
11
2003
XM_005252001.2
XP_005252058.1
10
1884
XM_005252003.2
XP_005252060.1
10
1700
XM_005252004.2
XP_005252061.1
9
1600
XM_005252006.3
XP_005252063.1
10
1884
XM_005252007.3
XP_005252064.1
10
1884
XM_011518718.2
XP_011517020.1
10
1884
XM_011518720.2
XP_011517022.1
10
1884
XM_017014751.1
XP_016870240.1
11
2426
XM_017014752.1
XP_016870241.1
11
1939
XM_017014753.1
XP_016870242.1
11
2426
XM_017014754.1
XP_016870243.1
9
1600
XM_017014755.1
XP_016870244.1
11
1939
XM_017014756.1
XP_016870245.1
10
1700
XM_017014757.1
XP_016870246.1
11
1939
XM_017014758.1
XP_016870247.1
11
1939
XM_017014759.1
XP_016870248.1
10
1700
XM_017014760.1
XP_016870249.1
11
2426
XM_017014761.1
XP_016870250.1
9
1600
GenBank mRNA
AB209118.1
-
11
1905
AF400441.1
-
8
1077
AF410899.1
-
9
1454
AF410900.1
-
9
1454
AF410901.1
-
9
1454
AF508964.1
-
8
1069
AK289904.1
-
9
1510
AK294285.1
-
8
845
BC031835.1
-
9
1381
4941763
S76473.1
-
8
1085
S76474.1
-
8
1085
U12140.1
-
8
1323
X75958.1
-
8
1069
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
neuron migration
positive regulation of protein phosphorylation
inflammatory response
neuromuscular junction development
aging
learning
long-term memory
circadian rhythm
feeding behavior
positive regulation of cell proliferation
response to light stimulus
positive regulation of gene expression
positive regulation of neuron projection development
response to auditory stimulus
glutamate secretion
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
central nervous system neuron development
cerebral cortex development
neuron differentiation
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
neurotrophin signaling pathway
mechanoreceptor differentiation
regulation of GTPase activity
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
retinal rod cell development
protein autophosphorylation
regulation of neurotransmitter secretion
oligodendrocyte differentiation
peripheral nervous system neuron development
positive regulation of axonogenesis
regulation of dendrite development
regulation of protein kinase B signaling
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
retina development in camera-type eye
long-term synaptic potentiation
cellular response to amino acid stimulus
cellular response to tumor necrosis factor
cellular response to brain-derived neurotrophic factor stimulus
negative regulation of anoikis
ATP binding
receptor tyrosine kinase binding
protein homodimerization activity
neurotrophin binding
brain-derived neurotrophic factor binding
brain-derived neurotrophic factor-activated receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-sshrn:80/10.239.165.44:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0