Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NUMA1 SIRNA Assays ›
Gene Symbol:
NUMA1
Gene Name:
nuclear mitotic apparatus protein 1
Gene Aliases:
NMP-22, NUMA
Chromosome Location:
Chr.11: 72002864 - 72080693 on Build GRCh38
UniGene ID:
Hs.325978
Species:
Human
siRNA ID
HSS107367
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NUMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286561.1
NP_001273490.1
11, 12
1175
NM_006185.3
NP_006176.2
10, 11
1077
NR_104476.1
10, 11
996
XM_006718564.1
XP_006718627.1
9, 10
892
XM_011545054.1
XP_011543356.1
9, 10
892
XM_011545055.1
XP_011543357.1
10, 11
1070
XM_011545056.2
XP_011543358.1
11, 12
1064
XM_011545057.2
XP_011543359.1
11, 12
1054
XM_011545058.1
XP_011543360.1
9, 10
1091
XM_011545059.2
XP_011543361.1
11, 12
1056
XM_011545060.2
XP_011543362.1
11, 12
1050
XM_011545061.2
XP_011543363.1
9, 10
910
XM_011545062.2
XP_011543364.1
9, 10
923
XM_011545063.2
XP_011543365.1
9, 10
890
XM_011545064.1
XP_011543366.1
9, 10
877
XM_011545065.1
XP_011543367.1
9, 10
892
XM_011545066.1
XP_011543368.1
9, 10
892
XM_017017830.1
XP_016873319.1
10, 11
932
XM_017017831.1
XP_016873320.1
9, 10
928
GenBank mRNA
AB210007.1
-
10, 11
878
AK290509.1
-
9, 10
892
AK307577.1
-
10, 11
895
AK309041.1
-
9, 10
899
BC004165.2
-
10, 11
911
BC027493.1
-
10, 11
878
BC043499.1
-
10, 11
878
BC103765.1
-
10, 11
996
Z11583.1
-
10, 11
895
Z11584.1
-
11, 12
997
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
nucleus organization
mitotic nuclear division
cell division
meiotic cell cycle
lung epithelial cell differentiation
structural molecule activity
protein binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-zz5lq:80/10.239.165.3:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0