Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018048.1 | NP_001018058.1 | 5 | 478 | ||
NM_001018049.2 | NP_001018059.1 | 5 | 544 | |||
NM_002571.3 | NP_002562.2 | 5 | 544 | |||
XM_011518745.2 | XP_011517047.1 | 5 | 616 | |||
XM_011518746.2 | XP_011517048.1 | 5 | 616 | |||
XM_011518747.2 | XP_011517049.1 | 5 | 616 | |||
XM_011518748.1 | XP_011517050.1 | 5 | 550 | |||
XM_011518749.2 | XP_011517051.1 | 5 | 544 | |||
XM_011518751.1 | XP_011517053.1 | 5 | 478 | |||
XM_011518752.2 | XP_011517054.1 | 4 | 433 | |||
XM_017014782.1 | XP_016870271.1 | 3 | 466 | |||
GenBank mRNA | AK304657.1 | - | 5 | 478 | ||
AK311813.1 | - | 5 | 544 | |||
BC069451.1 | - | 5 | 513 | 7262179 | ||
BC069562.1 | - | 5 | 513 | 7262096 | ||
BC112304.1 | - | 5 | 498 | 8327772 | ||
BC113728.1 | - | 5 | 498 | 8322780 | ||
DC421713.1 | - | 5 | 478 | |||
DQ884407.1 | - | 4 | 387 | |||
J04129.1 | - | 5 | 542 | |||
M61886.1 | - | 5 | 533 |