Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004562.2 | NP_004553.2 | 9 | 1179 | ||
NM_013987.2 | NP_054642.2 | 8 | 1095 | |||
NM_013988.2 | NP_054643.2 | 6 | 732 | |||
XM_011535863.1 | XP_011534165.1 | 9 | 1176 | |||
XM_017010908.1 | XP_016866397.1 | 9 | 2093 | |||
XM_017010909.1 | XP_016866398.1 | 8 | 973 | |||
GenBank mRNA | AB009973.1 | - | 9 | 1146 | ||
AB245403.1 | - | 10 | 1137 | |||
AF381282.1 | - | 7 | 804 | |||
AF381284.1 | - | 7 | 923 | |||
AF381286.1 | - | 2 | 181 | |||
AK292590.1 | - | 9 | 1140 | |||
AK294684.1 | - | 4 | 385 | |||
BC022014.2 | - | 10 | 1200 | 4824892 | ||
EF375726.1 | - | 8 | 1045 | |||
GU345837.1 | - | 8 | 898 | |||
GU345838.1 | - | 7 | 940 | |||
GU345839.1 | - | 8 | 978 | |||
GU357501.1 | - | 6 | 535 | |||
GU357502.1 | - | 4 | 473 | |||
GU361466.1 | - | 6 | 878 | |||
GU361469.1 | - | 9 | 1159 | |||
GU361470.1 | - | 9 | 1160 | |||
GU361471.1 | - | 3 | 234 | |||
KC357595.1 | - | 11 | 1257 | |||
KC774171.1 | - | 7 | 912 | |||
KJ534911.1 | - | 8 | 1045 | |||
KJ534912.1 | - | 8 | 1045 | |||
KU178240.1 | - | 9 | 1102 | |||
KU178241.1 | - | 1045 | ||||
KU178242.1 | - | 1042 |