Gene Symbol: PARN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134477.2 | NP_001127949.1 | 7 | 536 | ||
NM_001242992.1 | NP_001229921.1 | 6 | 481 | |||
NM_002582.3 | NP_002573.1 | 7 | 619 | |||
XM_011522510.2 | XP_011520812.1 | 7 | 625 | |||
XM_011522511.2 | XP_011520813.1 | 7 | 624 | |||
XM_011522513.2 | XP_011520815.1 | 7 | 535 | |||
XM_011522514.1 | XP_011520816.1 | 7 | 623 | |||
XM_017023258.1 | XP_016878747.1 | 7 | 623 | |||
XM_017023259.1 | XP_016878748.1 | 4 | 268 | |||
XM_017023260.1 | XP_016878749.1 | 2 | 157 | |||
GenBank mRNA | AJ005698.1 | - | 7 | 530 | ||
AK293189.1 | - | 7 | 528 | |||
AK301648.1 | - | 6 | 471 | |||
AK303348.1 | - | 7 | 515 | |||
AK315020.1 | - | 7 | 615 | |||
BC044763.1 | - | 7 | 557 | |||
BC050029.1 | - | 7 | 557 | 4825218 |