Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCCA SIRNA Assays ›
Gene Symbol:
PCCA
Gene Name:
propionyl-CoA carboxylase alpha subunit
Gene Aliases:
-
Chromosome Location:
Chr.13: 100089015 - 100530437 on Build GRCh38
UniGene ID:
Hs.733632
Species:
Human
siRNA ID
HSS107635
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCCA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000282.3
NP_000273.2
13
1225
NM_001127692.2
NP_001121164.1
12
1147
NM_001178004.1
NP_001171475.1
13
1225
XM_005254059.2
XP_005254116.1
13
1220
XM_011521093.1
XP_011519395.1
13
1220
XM_017020605.1
XP_016876094.1
13
1220
XM_017020606.1
XP_016876095.1
12
1071
XM_017020607.1
XP_016876096.1
13
1036
XM_017020608.1
XP_016876097.1
13
1220
XM_017020609.1
XP_016876098.1
13
1133
XM_017020610.1
XP_016876099.1
11
1487
XM_017020611.1
XP_016876100.1
13
1220
XM_017020612.1
XP_016876101.1
13
1220
XM_017020613.1
XP_016876102.1
13
1220
XM_017020614.1
XP_016876103.1
12
1071
XM_017020615.1
XP_016876104.1
13
1220
XM_017020616.1
XP_016876105.1
13
1220
XM_017020617.1
XP_016876106.1
9
804
XM_017020618.1
XP_016876107.1
9
804
GenBank mRNA
AF385926.1
-
13
1157
AK225288.1
-
13
1125
AK225299.1
-
13
1123
AK296771.1
-
12
1045
AK298318.1
-
13
1147
AK315503.1
-
13
1123
BC000140.2
-
13
1097
3048998
S55656.1
-
1
30
X14608.1
-
13
1092
Back To Top
More Information
Panther Classification:
Molecular Function -
ligase
Gene Ontology Categories:
Function(s)
Process(es)
biotin metabolic process
short-chain fatty acid catabolic process
biotin carboxylase activity
propionyl-CoA carboxylase activity
ATP binding
biotin binding
enzyme binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0