Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SERPINB6 SIRNA Assays ›
Gene Symbol:
SERPINB6
Gene Name:
serpin family B member 6
Gene Aliases:
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3
Chromosome Location:
Chr.6: 2948159 - 2972165 on Build GRCh38
UniGene ID:
Hs.519523
Species:
Human
siRNA ID
HSS107963
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SERPINB6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195291.2
NP_001182220.2
5
1134
NM_001271822.1
NP_001258751.1
4
559
NM_001271823.1
NP_001258752.1
4
657
NM_001271824.1
NP_001258753.1
4
1041
NM_001271825.1
NP_001258754.1
4
643
NM_001297699.1
NP_001284628.1
4
1284
NM_001297700.1
NP_001284629.1
4
940
NM_004568.5
NP_004559.4
4
999
XM_011514672.1
XP_011512974.1
6
937
XM_011514673.1
XP_011512975.1
4
1091
XM_011514674.2
XP_011512976.1
4
676
XM_011514676.2
XP_011512978.1
4
486
XM_017010940.1
XP_016866429.1
5
498
XM_017010941.1
XP_016866430.1
3
2127
GenBank mRNA
AB209563.1
-
3
667
AF116727.1
-
1
1220
AK057138.1
-
3
2358
AK094983.1
-
5
1134
AK098566.1
-
4
656
AK122895.1
-
2
1262
AK122961.1
-
3
1275
AK128637.1
-
2
2688
AK314578.1
-
4
718
BC001394.2
-
4
581
3051381
BC098564.1
-
4
437
30563105
BG499326.1
-
4
516
BI090002.1
-
4
641
BI259189.1
-
4
461
BM789272.1
-
4
622
BP325314.1
-
4
436
BP370484.1
-
4
492
BQ681973.1
-
4
446
BX375950.2
-
4
725
BX538343.1
-
4
4711
CX755337.1
-
4
562
CX782163.1
-
4
509
Z22658.1
-
4
430
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
sensory perception of sound
negative regulation of endopeptidase activity
cellular response to osmotic stress
protease binding
serine-type endopeptidase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0