Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006219.2 | NP_006210.1 | 2 | 236 | ||
XM_005247530.2 | XP_005247587.1 | 4 | 585 | |||
XM_006713659.3 | XP_006713722.1 | 3 | 456 | |||
XM_011512895.2 | XP_011511197.1 | 5 | 581 | |||
XM_017006619.1 | XP_016862108.1 | 3 | 259 | |||
GenBank mRNA | BC114432.1 | - | 2 | 224 | 40008544 | |
S67334.1 | - | 2 | 220 |