Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PMS1 SIRNA Assays ›
Gene Symbol:
PMS1
Gene Name:
PMS1 homolog 1, mismatch repair system component
Gene Aliases:
HNPCC3, MLH2, PMSL1, hPMS1
Chromosome Location:
Chr.2: 189784085 - 189877629 on Build GRCh38
UniGene ID:
Hs.111749
Species:
Human
siRNA ID
HSS108171
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000534.4
NP_000525.1
12
3066
NM_001128143.1
NP_001121615.1
11
2949
NM_001128144.1
NP_001121616.1
11
2580
NM_001289408.1
NP_001276337.1
11
2963
NM_001289409.1
NP_001276338.1
10
2780
NM_001321044.1
NP_001307973.1
10
2463
NM_001321045.1
NP_001307974.1
13
3192
NM_001321046.1
NP_001307975.1
11
2883
NM_001321047.1
NP_001307976.1
12
3243
NM_001321048.1
NP_001307977.1
13
3163
NR_110332.1
12
2792
XM_006712596.1
XP_006712659.1
8
2031
XM_011511356.2
XP_011509658.1
12
3352
XM_017004344.1
XP_016859833.1
12
2675
XM_017004345.1
XP_016859834.1
11
2500
XM_017004346.1
XP_016859835.1
11
2463
XM_017004347.1
XP_016859836.1
10
2366
XM_017004348.1
XP_016859837.1
11
2306
XM_017004349.1
XP_016859838.1
11
3235
XM_017004350.1
XP_016859839.1
10
2006
XM_017004351.1
XP_016859840.1
9
1968
GenBank mRNA
AB102870.1
-
10
2051
AB102872.1
-
6
1406
AB102875.1
-
10
2420
AK295602.1
-
11
2559
AK316215.1
-
10
2300
AY540750.1
-
7
1942
AY540751.1
-
12
2364
BC096330.1
-
12
2607
BC096331.1
-
11
2120
40008290
BC096332.1
-
12
2607
BG195153.1
-
1
70
BM553209.1
-
4
793
BM677668.1
-
2
469
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mismatch repair
DNA binding
single-stranded DNA binding
ATP binding
ATPase activity
mismatched DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0