Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU1F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000306.3 | NP_000297.1 | 2 | 288 | ||
NM_001122757.2 | NP_001116229.1 | 2 | 366 | |||
GenBank mRNA | D10216.1 | - | 2 | 288 | ||
L18781.1 | - | 2 | 163 | |||
X62429.1 | - | 2 | 182 | |||
X72215.1 | - | 2 | 163 |