Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU2F2 SIRNA Assays ›
Gene Symbol:
POU2F2
Gene Name:
POU class 2 homeobox 2
Gene Aliases:
OCT2, OTF2, Oct-2
Chromosome Location:
Chr.19: 42086110 - 42197931 on Build GRCh38
UniGene ID:
Hs.147381
Species:
Human
siRNA ID
HSS108274
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POU2F2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207025.2
NP_001193954.1
3
170
NM_001207026.1
NP_001193955.1
3
170
NM_001247994.1
NP_001234923.1
3
170
NM_002698.4
NP_002689.1
3
170
XM_005259010.3
XP_005259067.1
3
187
XM_011527040.2
XP_011525342.1
3
187
XM_011527041.2
XP_011525343.1
3
187
XM_011527042.2
XP_011525344.1
3
187
XM_011527043.2
XP_011525345.2
3
187
XM_017026884.1
XP_016882373.1
3
188
XM_017026885.1
XP_016882374.1
3
191
XM_017026886.1
XP_016882375.1
4
1690
XM_017026887.1
XP_016882376.1
4
160
XM_017026888.1
XP_016882377.1
4
640
XM_017026889.1
XP_016882378.1
3
182
XM_017026890.1
XP_016882379.1
3
263
XM_017026891.1
XP_016882380.1
3
188
XM_017026892.1
XP_016882381.1
3
188
XM_017026893.1
XP_016882382.1
3
187
XM_017026894.1
XP_016882383.1
3
187
XM_017026895.1
XP_016882384.1
3
185
XM_017026896.1
XP_016882385.1
3
188
GenBank mRNA
BC006101.1
-
3
162
3688152
BT007438.1
-
3
108
CB995118.1
-
3
203
M36542.1
-
3
162
M36653.1
-
3
174
M36718.1
-
3
105
X13809.1
-
3
105
X13810.1
-
3
162
X53468.1
-
3
174
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mature B cell differentiation
immunoglobulin secretion involved in immune response
transcription from RNA polymerase II promoter
humoral immune response
snRNA transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
cell maturation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein domain specific binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0