Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU3F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005604.3 | NP_005595.2 | 1 | 1209 | ||
GenBank mRNA | AK095588.1 | - | 3 | 1008 | ||
BC051699.1 | - | 1 | 1178 | 4817001 | ||
Z11933.1 | - | 1 | 1201 |