Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001928.2 | NP_001001928.1 | 4 | 509 | ||
NM_005036.4 | NP_005027.2 | 5 | 593 | |||
XM_005261655.3 | XP_005261712.1 | 5 | 886 | |||
XM_005261656.3 | XP_005261713.1 | 6 | 1050 | |||
XM_006724269.3 | XP_006724332.1 | 5 | 652 | |||
XM_006724270.3 | XP_006724333.1 | 4 | 558 | |||
XM_011530239.2 | XP_011528541.1 | 6 | 742 | |||
XM_011530240.2 | XP_011528542.1 | 6 | 745 | |||
XM_011530241.2 | XP_011528543.1 | 5 | 650 | |||
XM_011530242.2 | XP_011528544.1 | 4 | 472 | |||
XM_011530243.2 | XP_011528545.1 | 3 | 713 | |||
XM_011530244.2 | XP_011528546.1 | 6 | 745 | |||
XM_011530245.2 | XP_011528547.1 | 4 | 561 | |||
XM_017028839.1 | XP_016884328.1 | 7 | 920 | |||
XM_017028840.1 | XP_016884329.1 | 6 | 745 | |||
GenBank mRNA | AB307690.1 | - | 2 | 327 | ||
BC000052.2 | - | 4 | 537 | |||
CR456547.1 | - | 3 | 463 | |||
CR457435.1 | - | 2 | 327 | |||
EU395809.1 | - | 2 | 327 | |||
EU650667.1 | - | 2 | 327 | |||
HQ692862.1 | - | 2 | 327 | |||
L02932.1 | - | 4 | 543 | |||
S74349.1 | - | 3 | 450 | |||
Y07619.1 | - | 2 | 327 |