Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002709.2 | NP_002700.1 | 4 | 867 | ||
NM_206876.1 | NP_996759.1 | 5 | 662 | |||
GenBank mRNA | AF092905.1 | - | 6 | 640 | ||
AK295957.1 | - | 4 | 525 | |||
AK303682.1 | - | 4 | 526 | |||
AK312329.1 | - | 4 | 804 | |||
AK316532.1 | - | 4 | 488 | |||
BC002697.2 | - | 4 | 583 | 3609386 | ||
BC012045.1 | - | 4 | 583 | 3631338 | ||
BT019744.1 | - | 4 | 502 | |||
BX647295.1 | - | 2 | 345 | |||
BX647970.1 | - | 5 | 701 | |||
CD656807.1 | - | 5 | 629 | |||
CR542263.1 | - | 4 | 502 | |||
CR542285.1 | - | 4 | 502 | |||
EU668339.1 | - | 4 | 583 | |||
X80910.1 | - | 4 | 760 |