Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002709.2 | NP_002700.1 | 4, 5 | 895 | ||
NM_206876.1 | NP_996759.1 | 5, 6 | 690 | |||
GenBank mRNA | AF092905.1 | - | 6, 7 | 668 | ||
AK295957.1 | - | 4, 5 | 553 | |||
AK312329.1 | - | 4, 5 | 832 | |||
AK316532.1 | - | 4, 5 | 516 | |||
BC002697.2 | - | 4, 5 | 611 | 3609386 | ||
BC012045.1 | - | 4, 5 | 611 | 3631338 | ||
BT019744.1 | - | 4, 5 | 530 | |||
BX647295.1 | - | 2, 3 | 373 | |||
BX647970.1 | - | 5, 6 | 729 | |||
CR542263.1 | - | 4, 5 | 530 | |||
CR542285.1 | - | 4, 5 | 530 | |||
EU668339.1 | - | 4, 5 | 611 | |||
X80910.1 | - | 4, 5 | 788 |