Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 6 | 779 | ||
NM_001271900.1 | NP_001258829.1 | 7 | 973 | |||
NM_001271948.1 | NP_001258877.1 | 6 | 1306 | |||
NM_181674.2 | NP_858060.2 | 6 | 1104 | |||
NM_181675.3 | NP_858061.2 | 6 | 826 | |||
NM_181676.2 | NP_858062.1 | 5 | 910 | |||
NM_181677.2 | NP_858063.1 | 7 | 736 | |||
NM_181678.2 | NP_858064.1 | 7 | 749 | |||
NR_073526.1 | 5 | 1221 | ||||
NR_073527.1 | 6 | 858 | ||||
GenBank mRNA | AK056192.1 | - | 6 | 786 | ||
AK289717.1 | - | 6 | 755 | |||
AK294659.1 | - | 7 | 882 | |||
AK295347.1 | - | 6 | 750 | |||
AK314810.1 | - | 6 | 1103 | |||
AL540315.3 | - | 4 | 440 | |||
BC031790.1 | - | 6 | 994 | 4939981 | ||
BX647887.1 | - | 6 | 839 | |||
M64930.1 | - | 5 | 1032 |