Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP3CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243974.1 | NP_001230903.1 | 8 | 1363 | ||
NM_001243975.1 | NP_001230904.1 | 8 | 1363 | |||
NM_005605.4 | NP_005596.2 | 8 | 1363 | |||
XM_017013610.1 | XP_016869099.1 | 8 | 1156 | |||
XM_017013611.1 | XP_016869100.1 | 8 | 1363 | |||
GenBank mRNA | AK055013.1 | - | 8 | 1514 | ||
AK299415.1 | - | 8 | 1197 | |||
AY007249.1 | - | 8 | 970 | |||
BC004864.2 | - | 8 | 1250 | 3677098 | ||
S46622.1 | - | 8 | 1209 |