- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 4 | 560 | ||
NM_005040.3 | NP_005031.1 | 5 | 922 | |||
NM_199418.3 | NP_955450.2 | 6 | 985 | |||
XM_005274093.1 | XP_005274150.1 | 5 | 652 | |||
GenBank mRNA | AK091786.1 | - | 6 | 812 | ||
AK302386.1 | - | 4 | 511 | |||
AK312919.1 | - | 5 | 743 | |||
BC001500.2 | - | 5 | 740 | 3510081 | ||
L13977.1 | - | 5 | 750 |