Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321053.1 | NP_001307982.1 | 4 | 671 | ||
NM_001321054.1 | NP_001307983.1 | 2 | 308 | |||
NM_002727.3 | NP_002718.2 | 3 | 456 | |||
XM_017016392.1 | XP_016871881.1 | 4 | 1084 | |||
GenBank mRNA | AK311873.1 | - | 3 | 368 | ||
BC015516.1 | - | 3 | 366 | 3883161 | ||
BG057351.1 | - | 1 | 345 | |||
BG548665.1 | - | 3 | 336 | |||
BP335278.1 | - | 3 | 412 | |||
BP363056.1 | - | 3 | 413 | |||
BQ051861.1 | - | 2 | 219 | |||
CB110512.1 | - | 3 | 412 | |||
CD677686.1 | - | 1 | 42 | |||
J03223.1 | - | 3 | 357 | |||
X12765.1 | - | 2 | 226 | |||
X17042.1 | - | 3 | 350 |