Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207013.1 | NP_001193942.1 | 1 | 220 | ||
NM_002828.3 | NP_002819.2 | 1 | 220 | |||
NM_080422.2 | NP_536347.1 | 1 | 220 | |||
NM_080423.2 | NP_536348.1 | 1 | 220 | |||
XM_005258124.3 | XP_005258181.1 | 1 | 218 | |||
XM_005258125.3 | XP_005258182.1 | 1 | 218 | |||
XM_011525706.2 | XP_011524008.1 | 1 | 143 | |||
XM_017025884.1 | XP_016881373.1 | 1 | 236 | |||
XM_017025886.1 | XP_016881375.1 | 1 | 201 | |||
XM_017025887.1 | XP_016881376.1 | 1 | 152 | |||
XM_017025888.1 | XP_016881377.1 | 1 | 152 | |||
GenBank mRNA | AK225706.1 | - | 1 | 99 | ||
AK290649.1 | - | 1 | 102 | |||
AK292570.1 | - | 1 | 120 | |||
BC008244.1 | - | 1 | 63 | 3872164 | ||
BC016727.1 | - | 1 | 132 | 4081330 | ||
BG699440.1 | - | 1 | 225 | |||
M25393.1 | - | 1 | 86 |