Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN3 SIRNA Assays ›
Gene Symbol:
PTPN3
Gene Name:
protein tyrosine phosphatase, non-receptor type 3
Gene Aliases:
PTP-H1, PTPH1
Chromosome Location:
Chr.9: 109375694 - 109538395 on Build GRCh38
UniGene ID:
Hs.436429
Species:
Human
siRNA ID
HSS108822
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145368.1
NP_001138840.1
20
2038
NM_001145369.1
NP_001138841.1
16
4222
NM_001145370.1
NP_001138842.1
15
4087
NM_001145371.1
NP_001138843.1
11
1656
NM_001145372.1
NP_001138844.1
10
1521
NM_002829.3
NP_002820.3
21
2173
NR_026918.1
8
943
XM_006717197.3
XP_006717260.1
21
2282
XM_006717199.3
XP_006717262.1
21
2307
XM_006717201.3
XP_006717264.1
20
2114
XM_006717202.3
XP_006717265.1
18
1876
XM_006717203.3
XP_006717266.1
18
1835
XM_006717204.3
XP_006717267.1
21
2209
XM_011518888.2
XP_011517190.1
21
2243
XM_011518889.2
XP_011517191.1
21
2913
XM_017014955.1
XP_016870444.1
21
2265
XM_017014956.1
XP_016870445.1
19
1986
XM_017014957.1
XP_016870446.1
20
2074
GenBank mRNA
AK295875.1
-
19
1958
AK296265.1
-
10
1305
AK296388.1
-
12
1582
AK303608.1
-
8
943
AK304434.1
-
20
2038
AK308349.1
-
8
906
AK316066.1
-
20
2038
AK316118.1
-
10
1447
BC033716.1
-
3
199
BC126117.1
-
20
2083
8991833
BC143848.1
-
19
1948
9052363
BX648253.1
-
15
4088
BX648735.1
-
16
2657
CR749204.1
-
9
1100
DQ104439.1
-
20
2061
M64572.1
-
20
2084
S39392.1
-
13
1484
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of mitotic cell cycle
negative regulation of membrane protein ectodomain proteolysis
liver regeneration
regulation of membrane depolarization during action potential
regulation of sodium ion transmembrane transporter activity
phosphotyrosine binding
protein tyrosine phosphatase activity
protein binding
cytoskeletal protein binding
sodium channel regulator activity
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0