Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002830.3 | NP_002821.1 | 2 | 889 | ||
XM_011511560.2 | XP_011509862.1 | 2 | 773 | |||
XM_011511562.2 | XP_011509864.1 | 2 | 773 | |||
XM_017004596.1 | XP_016860085.1 | 2 | 773 | |||
XM_017004597.1 | XP_016860086.1 | 2 | 279 | |||
XM_017004598.1 | XP_016860087.1 | 2 | 773 | |||
XM_017004599.1 | XP_016860088.1 | 2 | 279 | |||
XM_017004600.1 | XP_016860089.1 | 2 | 279 | |||
XM_017004601.1 | XP_016860090.1 | 2 | 773 | |||
XM_017004602.1 | XP_016860091.1 | 2 | 773 | |||
XM_017004603.1 | XP_016860092.1 | 2 | 279 | |||
XM_017004604.1 | XP_016860093.1 | 2 | 773 | |||
XM_017004605.1 | XP_016860094.1 | 2 | 773 | |||
XM_017004606.1 | XP_016860095.1 | 2 | 773 | |||
XM_017004607.1 | XP_016860096.1 | 2 | 773 | |||
XM_017004608.1 | XP_016860097.1 | 2 | 773 | |||
GenBank mRNA | AK314836.1 | - | 2 | 701 | ||
BC010674.1 | - | 2 | 545 | 3853914 | ||
M68941.1 | - | 2 | 889 |