Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161440.2 | NP_001154912.2 | 18 | 2877 | ||
NM_002842.4 | NP_002833.4 | 20 | 3411 | |||
XM_011527183.1 | XP_011525485.1 | 20 | 3432 | |||
XM_011527188.1 | XP_011525490.1 | 19 | 3381 | |||
XM_017027056.1 | XP_016882545.1 | 19 | 3419 | |||
XM_017027057.1 | XP_016882546.1 | 19 | 3407 | |||
XM_017027058.1 | XP_016882547.1 | 19 | 3452 | |||
XM_017027059.1 | XP_016882548.1 | 19 | 3371 | |||
XM_017027060.1 | XP_016882549.1 | 18 | 3371 | |||
XM_017027061.1 | XP_016882550.1 | 17 | 2885 | |||
XM_017027062.1 | XP_016882551.1 | 19 | 3327 | |||
XM_017027063.1 | XP_016882552.1 | 19 | 3320 | |||
GenBank mRNA | AA573849.1 | - | 1 | 521 | ||
BC111715.1 | - | 19 | 2827 | 40008777 | ||
BC111716.1 | - | 20 | 3361 | 40008775 | ||
D15049.2 | - | 20 | 3385 |