Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
綜合式 CRO CDMO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRS SIRNA Assays ›
Gene Symbol:
PTPRS
Gene Name:
protein tyrosine phosphatase, receptor type S
Gene Aliases:
PTPSIGMA, R-PTP-S, R-PTP-sigma
Chromosome Location:
Chr.19: 5205503 - 5340803 on Build GRCh38
UniGene ID:
Hs.744928
Species:
Human
siRNA ID
HSS108884
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002850.3
NP_002841.3
20, 21
3688
NM_130853.2
NP_570923.2
12, 13
2395
NM_130854.2
NP_570924.2
16, 17
3622
NM_130855.2
NP_570925.2
13, 14
2407
XM_005259600.2
XP_005259657.1
17, 18
3617
XM_005259606.2
XP_005259663.1
16, 17
3590
XM_005259607.2
XP_005259664.1
16, 17
3590
XM_005259609.1
XP_005259666.1
12, 13
2362
XM_005259610.1
XP_005259667.1
12, 13
2362
XM_011528157.2
XP_011526459.1
11, 12
2233
XM_011528158.2
XP_011526460.1
5, 6
1096
XM_017027065.1
XP_016882554.1
18, 19
3629
XM_017027066.1
XP_016882555.1
18, 19
3647
XM_017027067.1
XP_016882556.1
18, 19
3545
XM_017027068.1
XP_016882557.1
18, 19
3629
XM_017027069.1
XP_016882558.1
17, 18
3602
XM_017027070.1
XP_016882559.1
18, 19
3629
XM_017027071.1
XP_016882560.1
18, 19
3629
XM_017027072.1
XP_016882561.1
17, 18
3602
XM_017027073.1
XP_016882562.1
12, 13
2832
XM_017027074.1
XP_016882563.1
13, 14
2374
XM_017027075.1
XP_016882564.1
13, 14
2374
XM_017027076.1
XP_016882565.1
13, 14
2374
GenBank mRNA
AB209333.1
-
12, 13
2395
BC104812.1
-
11, 12
2235
8143815
BC143286.1
-
11, 12
2235
BC143287.1
-
11, 12
2235
9051790
U35234.1
-
19, 20
3562
U40317.1
-
15, 16
3413
U41725.1
-
11, 12
2186
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
chemical synaptic transmission
spinal cord development
cerebellum development
hippocampus development
cerebral cortex development
corpus callosum development
peptidyl-tyrosine dephosphorylation
establishment of endothelial intestinal barrier
transmembrane receptor protein tyrosine phosphatase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0