Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PEX5 SIRNA Assays ›
Gene Symbol:
PEX5
Gene Name:
peroxisomal biogenesis factor 5
Gene Aliases:
PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
Chromosome Location:
Chr.12: 7188072 - 7218574 on Build GRCh38
UniGene ID:
Hs.567327
Species:
Human
siRNA ID
HSS108930
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PEX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000319.4
NP_000310.2
8
784
NM_001131023.1
NP_001124495.1
9
894
NM_001131024.1
NP_001124496.1
8
738
NM_001131025.1
NP_001124497.1
9
1315
NM_001131026.1
NP_001124498.1
10
1348
NM_001300789.1
NP_001287718.1
9
920
XM_005253455.1
XP_005253512.1
8
804
XM_011520793.2
XP_011519095.1
9
909
XM_011520795.1
XP_011519097.1
9
1326
XM_011520797.1
XP_011519099.1
9
819
XM_011520798.1
XP_011519100.1
10
1028
XM_011520799.2
XP_011519101.1
9
1468
XM_011520800.1
XP_011519102.1
9
914
XM_011520801.2
XP_011519103.1
8
797
XM_011520802.2
XP_011519104.1
8
671
XM_017019744.1
XP_016875233.1
8
1253
XM_017019745.1
XP_016875234.1
8
1254
XM_017019746.1
XP_016875235.1
7
1142
XM_017019747.1
XP_016875236.1
7
1145
XM_017019748.1
XP_016875237.1
9
1517
XM_017019749.1
XP_016875238.1
9
1281
XM_017019750.1
XP_016875239.1
9
819
XM_017019751.1
XP_016875240.1
8
819
XM_017019752.1
XP_016875241.1
9
917
XM_017019753.1
XP_016875242.1
8
803
XM_017019754.1
XP_016875243.1
8
1014
XM_017019755.1
XP_016875244.1
8
1412
XM_017019756.1
XP_016875245.1
8
708
XM_017019757.1
XP_016875246.1
9
917
GenBank mRNA
AK225126.1
-
8
693
AK292256.1
-
10
1348
AK299105.1
-
8
738
AK299534.1
-
4
249
AK301700.1
-
5
454
AK302742.1
-
9
854
AK303515.1
-
9
920
AK312317.1
-
9
1253
AK316250.1
-
8
693
BC010621.2
-
9
807
3872822
KU178292.1
-
8
768
KU178293.1
-
8
768
KU178294.1
-
7
657
U19721.1
-
7
671
X84899.1
-
8
786
Z48054.1
-
8
743
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into peroxisome matrix
protein import into peroxisome matrix, docking
protein import into peroxisome membrane
negative regulation of protein homotetramerization
peroxisome targeting sequence binding
peroxisome matrix targeting signal-1 binding
protein binding
enzyme binding
small GTPase binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-b8dk7:80/10.239.160.22:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0