Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PYGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164716.1 | NP_001158188.1 | 16, 17 | 2873 | ||
GenBank mRNA | AF066859.1 | - | 18, 19 | 2320 | ||
AK056607.1 | - | 16, 17 | 2132 | |||
AK057547.1 | - | 1, 2 | 1672 | |||
AK299376.1 | - | 14, 15 | 1660 | |||
AK299461.1 | - | 18, 19 | 2339 | |||
AK314474.1 | - | 18, 19 | 2324 | |||
BC126392.1 | - | 18, 19 | 2719 | 8992108 | ||
BC130514.1 | - | 18, 19 | 2719 | 40146545 | ||
X03031.1 | - | 2, 3 | 297 |