Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243738.1 | NP_001230667.1 | 10, 11 | 1718 | ||
NM_004761.4 | NP_004752.1 | 11, 12 | 1915 | |||
XM_011514785.2 | XP_011513087.1 | 9, 10 | 1187 | |||
GenBank mRNA | AB012295.1 | - | 10, 11 | 1267 | ||
AK095967.1 | - | 8, 9 | 889 | |||
AK294442.1 | - | 10, 11 | 1254 | |||
AL050259.1 | - | 11, 12 | 1488 | |||
BC000316.1 | - | 5, 6 | 574 | |||
BC005037.1 | - | 8, 9 | 956 | |||
BC032681.1 | - | 11, 12 | 1470 | 5585507 | ||
BC040989.1 | - | 10, 11 | 1770 | |||
BX648015.1 | - | 10, 11 | 1260 | |||
U68142.1 | - | 5, 6 | 521 |