Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004582.3 | NP_004573.2 | 7, 8 | 772 | ||
NR_073562.1 | 9, 10 | 967 | ||||
GenBank mRNA | AB209023.1 | - | 6, 7 | 694 | ||
AK128861.1 | - | 5, 6 | 3375 | |||
AK293057.1 | - | 9, 10 | 967 | |||
BC020790.1 | - | 7, 8 | 765 | 4290610 | ||
CR456941.1 | - | 6, 7 | 694 | |||
U49245.1 | - | 6, 7 | 694 | |||
X98001.1 | - | 6, 7 | 718 | |||
Y08201.1 | - | 6, 7 | 697 |