Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270362.1 | NP_001257291.1 | 8 | 969 | ||
NM_005053.3 | NP_005044.1 | 8 | 972 | |||
NR_072976.1 | 7 | 916 | ||||
GenBank mRNA | AB209713.1 | - | 8 | 892 | ||
AK289908.1 | - | 8 | 906 | |||
AK293219.1 | - | 7 | 875 | |||
BC014026.2 | - | 8 | 898 | 4300551 | ||
BC088364.1 | - | 8 | 898 | 30389208 | ||
D21235.1 | - | 8 | 873 | |||
M77024.1 | - | 4 | 284 |