Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002892.3 | NP_002883.3 | 15 | 1455 | ||
NM_023000.2 | NP_075376.2 | 15 | 1455 | |||
NM_023001.2 | NP_075377.2 | 15 | 1455 | |||
XM_005267964.2 | XP_005268021.1 | 15 | 1574 | |||
XM_005267965.2 | XP_005268022.1 | 15 | 1574 | |||
XM_017021560.1 | XP_016877049.1 | 15 | 1574 | |||
XM_017021561.1 | XP_016877050.1 | 15 | 1351 | |||
XM_017021562.1 | XP_016877051.1 | 15 | 1574 | |||
XM_017021563.1 | XP_016877052.1 | 15 | 1574 | |||
XM_017021564.1 | XP_016877053.1 | 13 | 1182 | |||
XM_017021565.1 | XP_016877054.1 | 5 | 1617 | |||
GenBank mRNA | AF086075.1 | - | 3 | 202 | ||
AK307520.1 | - | 15 | 1404 | |||
AK307972.1 | - | 433 | ||||
BC026230.1 | - | 15 | 1403 | |||
S57153.1 | - | 3 | 190 | |||
S57160.1 | - | 3 | 190 | |||
S57162.1 | - | 3 | 190 | |||
S66427.1 | - | 15 | 1315 |