Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006743.4 | NP_006734.1 | 5 | 571 | ||
GenBank mRNA | AK000859.1 | - | 5 | 440 | ||
AK026664.1 | - | 2 | 543 | |||
AK309137.1 | - | 5 | 829 | |||
AY203954.1 | - | 4 | 1033 | |||
BC006825.1 | - | 5 | 400 | 3449185 | ||
BI523929.1 | - | 5 | 517 | |||
BQ429805.1 | - | 4 | 335 | |||
BX647914.1 | - | 1 | 2015 | |||
CD559009.1 | - | 5 | 628 |