Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199771.1 | NP_001186700.1 | 5 | 942 | ||
NM_002905.3 | NP_002896.2 | 5 | 937 | |||
GenBank mRNA | BC028298.1 | - | 5 | 896 | 4942311 | |
BU617433.1 | - | 1 | 414 | |||
BU731044.1 | - | 1 | 376 | |||
U43559.1 | - | 5 | 830 | |||
U89717.1 | - | 5 | 867 |
Gene Symbol: BLOC1S1-RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037658.1 | 6 | 813 | |||
GenBank mRNA | AK096656.1 | - | 4 | 3004 |
Gene Symbol: CPT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309464.1 | - | 2071 |