Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002929.2 | NP_002920.1 | 5 | 1261 | ||
XM_017020684.1 | XP_016876173.1 | 5 | 2928 | |||
GenBank mRNA | AK289912.1 | - | 5 | 1385 | ||
BC140945.1 | - | 5 | 1226 | 9056757 | ||
L77503.1 | - | 5 | 1168 | |||
U63973.1 | - | 5 | 1261 |