Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000326.4 | NP_000317.1 | 8 | 1120 | ||
XM_011521870.2 | XP_011520172.1 | 8 | 2549 | |||
XM_017022460.1 | XP_016877949.1 | 5 | 959 | |||
GenBank mRNA | AK312457.1 | - | 8 | 1008 | ||
BC004199.2 | - | 8 | 1022 | 3604048 | ||
BG205561.1 | - | 2 | 215 | |||
BM704704.1 | - | 2 | 127 | |||
J04213.1 | - | 7 | 923 |