Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005977.3 | NP_005968.1 | 4 | 739 | ||
NM_183043.2 | NP_898864.1 | 4 | 480 | |||
NM_183044.2 | NP_898865.1 | 4 | 614 | |||
NM_183045.1 | NP_898866.1 | 4 | 739 | |||
XM_005266485.2 | XP_005266542.1 | 4 | 1085 | |||
XM_005266486.2 | XP_005266543.1 | 4 | 533 | |||
XM_011535177.2 | XP_011533479.1 | 4 | 1107 | |||
XM_011535178.1 | XP_011533480.1 | 4 | 1138 | |||
XM_017020685.1 | XP_016876174.1 | 4 | 1138 | |||
XM_017020686.1 | XP_016876175.1 | 4 | 689 | |||
XM_017020687.1 | XP_016876176.1 | 4 | 533 | |||
GenBank mRNA | AF293342.1 | - | 4 | 629 | ||
AJ010346.1 | - | 4 | 629 | |||
AJ010347.1 | - | 4 | 430 | |||
AK293272.1 | - | 4 | 401 | |||
AK312435.1 | - | 4 | 684 | |||
AY009109.1 | - | 4 | 562 | |||
BC034688.2 | - | 3 | 380 | 4508997 | ||
BP310130.1 | - | 4 | 553 | |||
CR749238.1 | - | 4 | 671 | |||
DC310579.1 | - | 4 | 480 | |||
KU178309.1 | - | 2 | 269 | |||
KU178310.1 | - | 2 | 269 |