Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319182.1 | NP_001306111.1 | 10 | 1516 | ||
NM_001319183.1 | NP_001306112.1 | 9 | 1802 | |||
NM_001319184.1 | NP_001306113.1 | 9 | 1656 | |||
NM_020216.3 | NP_064601.3 | 10 | 1805 | |||
XM_005245421.1 | XP_005245478.1 | 9 | 1323 | |||
XM_017002022.1 | XP_016857511.1 | 9 | 1490 | |||
GenBank mRNA | AJ242586.1 | - | 10 | 1806 | ||
AJ296161.1 | - | 10 | 1776 | |||
AK026644.1 | - | 7 | 1008 | |||
AK097372.1 | - | 11 | 1719 | |||
AL390139.1 | - | 9 | 1485 | |||
BC001064.2 | - | 10 | 1790 | 2823082 | ||
BC012166.1 | - | 10 | 1782 | 4636934 | ||
BX648806.1 | - | 1 | 724 |