Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319182.1 | NP_001306111.1 | 3 | 376 | ||
NM_020216.3 | NP_064601.3 | 3 | 665 | |||
XM_017002022.1 | XP_016857511.1 | 2 | 204 | |||
GenBank mRNA | AJ296161.1 | - | 3 | 636 | ||
AK097372.1 | - | 4 | 579 | |||
BC001064.2 | - | 3 | 650 | 2823082 | ||
BC012166.1 | - | 3 | 642 | 4636934 | ||
BG388104.1 | - | 3 | 376 | |||
BQ898593.1 | - | 665 | ||||
BY994108.1 | - | 3 | 353 | |||
DA684772.1 | - | 476 |