Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195728.2 | NP_001182657.1 | 8 | 1152 | ||
NM_001252652.1 | NP_001239581.1 | 8 | 1217 | |||
NM_004171.3 | NP_004162.2 | 7 | 1514 | |||
XM_005253067.1 | XP_005253124.1 | 7 | 991 | |||
XM_011520285.2 | XP_011518587.1 | 7 | 1028 | |||
XM_017018136.1 | XP_016873625.1 | 7 | 1811 | |||
XM_017018137.1 | XP_016873626.1 | 7 | 1031 | |||
XM_017018138.1 | XP_016873627.1 | 7 | 1015 | |||
GenBank mRNA | AB209444.1 | - | 7 | 930 | ||
AK122843.1 | - | 8 | 1217 | |||
AK298769.1 | - | 7 | 1026 | |||
AY066021.1 | - | 7 | 1205 | |||
BC132768.1 | - | 7 | 1081 | 40146790 | ||
D85884.1 | - | 7 | 999 | |||
KP966086.1 | - | 7 | 922 | |||
U01824.1 | - | 8 | 1073 | |||
U03505.1 | - | 7 | 955 | |||
Z32517.1 | - | 7 | 1011 |