Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001046.2 | NP_001037.1 | 10 | 1842 | ||
NM_001256461.1 | NP_001243390.1 | 10 | 1842 | |||
NR_046207.1 | 10 | 1842 | ||||
XM_011543588.2 | XP_011541890.1 | 10 | 1836 | |||
XM_017009771.1 | XP_016865260.1 | 9 | 992 | |||
GenBank mRNA | AY280459.1 | - | 10 | 1867 | ||
BC144221.1 | - | 10 | 1785 | 9052741 | ||
BC146839.1 | - | 10 | 1785 | 9056767 | ||
U30246.1 | - | 10 | 1842 |