Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145961.1 | NP_001139433.1 | 10 | 1511 | ||
NM_001145962.1 | NP_001139434.1 | 9 | 1416 | |||
NM_001145963.1 | NP_001139435.1 | 10 | 1390 | |||
NM_001145964.1 | NP_001139436.1 | 10 | 1381 | |||
NM_005072.4 | NP_005063.1 | 10 | 1511 | |||
GenBank mRNA | AF047338.1 | - | 10 | 1370 | ||
AF053402.1 | - | 4 | 533 | |||
AF054505.1 | - | 10 | 1370 | |||
AF054506.1 | - | 10 | 1370 | |||
AK026911.1 | - | 2 | 149 | |||
AK293906.1 | - | 10 | 1420 | |||
AK293956.1 | - | 9 | 1306 | |||
AK299042.1 | - | 10 | 1297 | |||
AK302790.1 | - | 10 | 1390 | |||
AK316415.1 | - | 10 | 1371 | |||
BC021193.2 | - | 10 | 1439 | 3349710 | ||
BC035480.1 | - | 10 | 2691 | |||
U55054.1 | - | 10 | 1425 |