Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324400.1 | NP_001311329.1 | 3 | 391 | ||
NM_022444.3 | NP_071889.2 | 2 | 235 | |||
XM_011516515.2 | XP_011514817.1 | 2 | 239 | |||
XM_011516516.2 | XP_011514818.1 | 2 | 239 | |||
XM_011516517.2 | XP_011514819.1 | 3 | 358 | |||
XM_011516518.2 | XP_011514820.1 | 2 | 239 | |||
XM_011516519.2 | XP_011514821.1 | 2 | 239 | |||
XM_017012555.1 | XP_016868044.1 | 2 | 239 | |||
GenBank mRNA | AF260824.1 | - | 2 | 235 | ||
AK026413.1 | - | 2 | 233 | |||
AK314537.1 | - | 2 | 233 | |||
BC111775.1 | - | 3 | 391 |