Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003057.2 | NP_003048.1 | 7 | 1251 | ||
NM_153187.1 | NP_694857.1 | 7 | 1251 | |||
XM_005267102.4 | XP_005267159.1 | 7 | 1293 | |||
XM_005267103.1 | XP_005267160.1 | 7 | 1293 | |||
XM_005267104.4 | XP_005267161.1 | 7 | 873 | |||
XM_005267105.4 | XP_005267162.1 | 7 | 825 | |||
XM_006715552.1 | XP_006715615.1 | 7 | 1293 | |||
XM_011536074.2 | XP_011534376.1 | 7 | 1116 | |||
GenBank mRNA | AK289887.1 | - | 7 | 1209 | ||
BC126364.1 | - | 7 | 1217 | 8992080 | ||
U77086.1 | - | 7 | 1199 | |||
X98332.1 | - | 7 | 1218 |