Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271946.1 | NP_001258875.1 | 20, 21 | 2691 | ||
NM_003062.3 | NP_003053.1 | 20, 21 | 2691 | |||
XM_017009779.1 | XP_016865268.1 | 20, 21 | 2094 | |||
GenBank mRNA | AB011538.2 | - | 20, 21 | 2691 | ||
AB017169.1 | - | 20, 21 | 2534 | |||
AF075240.1 | - | 4, 5 | 552 | |||
AY358884.1 | - | 20, 21 | 2353 | |||
BC146759.1 | - | 20, 21 | 2691 | 9007197 |