Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SNAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322902.1 | NP_001309831.1 | 7, 8 | 827 | ||
NM_001322903.1 | NP_001309832.1 | 8, 9 | 923 | |||
NM_001322904.1 | NP_001309833.1 | 8, 9 | 893 | |||
NM_001322905.1 | NP_001309834.1 | 8, 9 | 856 | |||
NM_001322906.1 | NP_001309835.1 | 8, 9 | 850 | |||
NM_001322907.1 | NP_001309836.1 | 9, 10 | 1051 | |||
NM_001322908.1 | NP_001309837.1 | 9, 10 | 974 | |||
NM_001322909.1 | NP_001309838.1 | 8, 9 | 841 | |||
NM_001322910.1 | NP_001309839.1 | 8, 9 | 948 | |||
NM_003081.4 | NP_003072.2 | 7, 8 | 796 | |||
NM_130811.3 | NP_570824.1 | 7, 8 | 796 | |||
XM_005260808.4 | XP_005260865.1 | 7, 8 | 874 | |||
XM_017028021.1 | XP_016883510.1 | 8, 9 | 726 | |||
XM_017028022.1 | XP_016883511.1 | 8, 9 | 818 | |||
GenBank mRNA | AK026732.1 | - | 2, 3 | 967 | ||
AK090857.1 | - | 2, 3 | 1764 | |||
AK094560.1 | - | 8, 9 | 882 | |||
AK098347.1 | - | 8, 9 | 524 | |||
AK098767.1 | - | 7, 8 | 666 | |||
AK223617.1 | - | 7, 8 | 758 | |||
AK289647.1 | - | 9, 10 | 957 | |||
AK314359.1 | - | 7, 8 | 707 | |||
BC010647.1 | - | 7, 8 | 685 | 3867544 | ||
BT019684.1 | - | 6, 7 | 553 | |||
BX647443.1 | - | 7, 8 | 668 | |||
D21267.1 | - | 7, 8 | 758 | |||
L19760.1 | - | 7, 8 | 641 | |||
L19761.1 | - | 7, 8 | 641 |